Assay Details
Target Gene Details
Entrez Gene ID: | 72 |
Gene Name: | actin, gamma 2, smooth muscle, enteric |
Gene Aliases: |
ACT, ACTA3, ACTE, ACTL3, ACTSG, VSCM |
Location: |
Chr.2:73892966-73919653 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTG2 | NM_001199893.1 | NP_001186822.1 | ||
| NM_001615.3 | NP_001606.1 | |||
| AK124338.1 | ||||
| AK304523.1 | ||||
| AK312955.1 | ||||
| BC012617.1 | AAH12617.1 | |||
| BC094877.1 | AAH94877.1 | |||
| BI850192.1 | ||||
| CR536515.1 | CAG38753.1 | |||
| CR541794.1 | CAG46593.1 | |||
| X16940.1 | CAA34814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010401 | Chr.2:73901234 - 73923975 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map