Assay Details
Target Gene Details
Entrez Gene ID: | 23550 |
Gene Name: | pleckstrin and Sec7 domain containing 4 |
Gene Aliases: |
EFA6B, TIC |
Location: |
Chr.2:113157399-113203229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSD4 | XM_005263634.2 | XP_005263691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582711 | Chr.2:113137939 - 113226926 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73955169] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map