Assay Details
Target Gene Details
Entrez Gene ID: | 3069 |
Gene Name: | high density lipoprotein binding protein |
Gene Aliases: |
HBP, PRO2900, VGL |
Location: |
Chr.2:241227264-241315842 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDLBP | NM_001320965.1 | NP_001307894.1 | ||
| NM_005336.5 | NP_005327.1 | |||
| XM_005247002.3 | XP_005247059.2 | |||
| XM_011511058.2 | XP_011509360.1 | |||
| XM_011511060.2 | XP_011509362.1 | |||
| XM_017003941.1 | XP_016859430.1 | |||
| XM_017003942.1 | XP_016859431.1 | |||
| AK296684.1 | ||||
| AK312335.1 | ||||
| BC001179.1 | AAH01179.1 | |||
| BG720303.1 | ||||
| BX647759.1 | ||||
| DC400438.1 | ||||
| DC424146.1 |
Target Gene Details
Entrez Gene ID: | 101927509 |
Gene Name: | basic proline-rich protein-like |
Gene Aliases: |
- |
Location: |
Chr.2:241267602-241277002 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927509 | XM_017005451.1 | 6 | 2551 | XP_016860940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594823 | Chr.2:241262775 - 241279993 on Build GRCh38 | Gain |
|
| nsv584957 | Chr.2:241257758 - 241312124 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map