Assay Details
Target Gene Details
Entrez Gene ID: | 3760 |
Gene Name: | potassium voltage-gated channel subfamily J member 3 |
Gene Aliases: |
GIRK1, KGA, KIR3.1 |
Location: |
Chr.2:154698581-154858352 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNJ3 | NM_001260508.1 | NP_001247437.1 | ||
| NM_002239.3 | NP_002230.1 | |||
| AK293824.1 | ||||
| AK293924.1 | ||||
| BC022495.1 | AAH22495.1 | |||
| GU074515.1 | ||||
| U50964.1 | AAB42176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834429 | Chr.2:154663084 - 154822774 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116080468] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map