Assay Details
Target Gene Details
Entrez Gene ID: | 150709 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
- |
Location: |
Chr.2:189674292-189763059 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 150709 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
- |
Location: |
Chr.2:189674292-189763059 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |