Assay Details
Target Gene Details
Entrez Gene ID: | 100506286 |
Gene Name: | uncharacterized LOC100506286 |
Gene Aliases: |
- |
Location: |
Chr.2:101151660-101155412 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506286 | NR_135594.1 | 1 | 457 | |
| AK056547.1 | 1 | 457 | ||
| DA744225.1 | 1 | 457 |
Target Gene Details
Entrez Gene ID: | 11138 |
Gene Name: | TBC1 domain family member 8 |
Gene Aliases: |
AD3, GRAMD8, HBLP1, TBC1D8A, VRP |
Location: |
Chr.2:101006435-101195326 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D8 | XM_011510507.1 | XP_011508809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834317 | Chr.2:101055657 - 101215215 on Build GRCh38 | Loss |
|
| esv34123 | Chr.2:101008454 - 101391851 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map