Assay Details
Target Gene Details
Entrez Gene ID: | 2181 |
Gene Name: | acyl-CoA synthetase long-chain family member 3 |
Gene Aliases: |
ACS3, FACL3, PRO2194 |
Location: |
Chr.2:222860984-222944624 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSL3 | NM_004457.3 | NP_004448.2 | ||
| NM_203372.1 | NP_976251.1 | |||
| XM_017003584.1 | XP_016859073.1 | |||
| AB061712.1 | BAB72074.1 | |||
| AK314621.1 | ||||
| BC041692.1 | AAH41692.1 | |||
| BG772246.1 | ||||
| BQ435549.1 | ||||
| BX472576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011830 | Chr.2:222673597 - 222979600 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map