Assay Details
Target Gene Details
Entrez Gene ID: | 10438 |
Gene Name: | C1D nuclear receptor corepressor |
Gene Aliases: |
LRP1, Rrp47, SUN-CoR, SUNCOR, hC1D |
Location: |
Chr.2:68042200-68063027 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1D | NM_001190263.1 | NP_001177192.1 | ||
| NM_001190265.1 | NP_001177194.1 | |||
| NM_006333.3 | NP_006324.1 | |||
| NM_173177.2 | NP_775269.1 | |||
| AK290451.1 | ||||
| BC005235.1 | AAH05235.1 | |||
| BC009584.1 | AAH09584.1 | |||
| BC016284.2 | AAH16284.1 | |||
| BG505686.1 | ||||
| CN342947.1 | ||||
| DA900336.1 | ||||
| X95592.1 | CAA64845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525802 | Chr.2:68033883 - 68051911 on Build GRCh38 | Loss |
|
| nsv834251 | Chr.2:68015596 - 68200603 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map