Assay Details
Target Gene Details
Entrez Gene ID: | 3557 |
Gene Name: | interleukin 1 receptor antagonist |
Gene Aliases: |
DIRA, ICIL-1RA, IL-1RN, IL-1ra, IL-1ra3, IL1F3, IL1RA, IRAP, MVCD4 |
Location: |
Chr.2:113099365-113134016 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RN | NM_000577.4 | NP_000568.1 | ||
| NM_001318914.1 | NP_001305843.1 | |||
| NM_173841.2 | NP_776213.1 | |||
| NM_173843.2 | NP_776215.1 | |||
| XM_011511121.1 | XP_011509423.1 | |||
| AF043143.1 | AAC39672.1 | |||
| AK290898.1 | ||||
| BC009745.2 | AAH09745.1 | |||
| DA667440.1 | ||||
| M55646.1 | AAA59138.1 | |||
| X84348.1 | CAA59087.1 |
More Information
Set Membership: |
|

Genomic Map