Assay Details
Target Gene Details
Entrez Gene ID: | 92 |
Gene Name: | activin A receptor type 2A |
Gene Aliases: |
ACTRII, ACVR2 |
Location: |
Chr.2:147844517-147930827 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR2A | NM_001278579.1 | NP_001265508.1 | ||
| NM_001278580.1 | NP_001265509.1 | |||
| NM_001616.4 | NP_001607.1 | |||
| AK301629.1 | ||||
| AK314124.1 | ||||
| BC067417.1 | AAH67417.1 | |||
| BC067418.1 | AAH67418.1 | |||
| BC069707.1 | AAH69707.1 | |||
| D31770.1 | BAA06548.1 | |||
| M93415.1 | AAA35504.1 | |||
| X62381.1 | CAA44245.1 | |||
| X63128.1 | CAA44839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955261 | Chr.2:147846632 - 147862231 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map