Assay Details
Target Gene Details
Entrez Gene ID: | 130340 |
Gene Name: | adaptor related protein complex 1 sigma 3 subunit |
Gene Aliases: |
PSORS15 |
Location: |
Chr.2:223755330-223837602 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1S3 | NM_001039569.1 | NP_001034658.1 | ||
| NR_110905.1 | ||||
| NR_110906.1 | ||||
| XM_011510600.2 | XP_011508902.1 | |||
| AK025338.1 | ||||
| AK299026.1 | ||||
| BC009606.1 | AAH09606.1 | |||
| BG340480.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834554 | Chr.2:223627491 - 223813918 on Build GRCh38 | Loss |
|
| nsv834555 | Chr.2:223728131 - 223887704 on Build GRCh38 | Loss |
|
| esv2763107 | Chr.2:223755950 - 223760359 on Build GRCh38 | Loss |
|
| nsv584549 | Chr.2:223436179 - 223828699 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map