Assay Details
Target Gene Details
Entrez Gene ID: | 805 |
Gene Name: | calmodulin 2 |
Gene Aliases: |
CAMII, LQT15, PHKD, PHKD2, caM |
Location: |
Chr.2:47160082-47176936 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALM2 | NM_001305624.1 | NP_001292553.1 | ||
| NM_001305625.1 | NP_001292554.1 | |||
| NM_001305626.1 | NP_001292555.1 | |||
| NM_001743.5 | NP_001734.1 | |||
| AK055130.1 | ||||
| AK315694.1 | ||||
| BC003354.1 | AAH03354.1 | |||
| BC006464.1 | AAH06464.1 | |||
| BC008437.1 | AAH08437.1 | |||
| BC017385.1 | AAH17385.1 | |||
| BC018677.1 | AAH18677.1 | |||
| BC026065.1 | AAH26065.1 | |||
| BG563041.1 | ||||
| BP370358.1 | ||||
| BT009916.1 | AAP88918.1 | |||
| CR541990.1 | CAG46787.1 | |||
| CR542021.1 | CAG46818.1 | |||
| D45887.1 | BAA08302.1 | |||
| HY022313.1 | ||||
| HY108355.1 | ||||
| M19311.1 | AAA35641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004828 | Chr.2:46829710 - 47587343 on Build GRCh38 | Gain |
|
| nsv581754 | Chr.2:47086485 - 47221726 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map