Assay Details
Target Gene Details
Entrez Gene ID: | 150864 |
Gene Name: | family with sequence similarity 117 member B |
Gene Aliases: |
ALS2CR13 |
Location: |
Chr.2:202635178-202769757 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM117B | NM_173511.3 | 1 | 121 | NP_775782.2 |
| CN397358.1 | 1 | 121 |
More Information
Set Membership: |
|

Genomic Map