Assay Details
Target Gene Details
Entrez Gene ID: | 389072 |
Gene Name: | pleckstrin homology domain containing M3 |
Gene Aliases: |
DAPR, PLEKHM1L |
Location: |
Chr.2:207821288-208025675 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHM3 | NM_001080475.2 | 8 | 5738 | NP_001073944.1 |
| XM_005246545.3 | 7 | 5580 | XP_005246602.1 | |
| XM_011511160.2 | 9 | 6073 | XP_011509462.1 | |
| XM_017004072.1 | 7 | 5923 | XP_016859561.1 | |
| XM_017004073.1 | 7 | 4802 | XP_016859562.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs147194338] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map