Assay Details
Target Gene Details
Entrez Gene ID: | 30845 |
Gene Name: | EH domain containing 3 |
Gene Aliases: |
PAST3 |
Location: |
Chr.2:31234014-31269451 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EHD3 | NM_014600.2 | NP_055415.1 | ||
| AF181264.1 | AAF40471.1 | |||
| AF214736.1 | AAF32285.1 | |||
| AK294222.1 | ||||
| AK316006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1128789 | Chr.2:31250334 - 31252634 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map