Assay Details
Target Gene Details
Entrez Gene ID: | 64240 |
Gene Name: | ATP binding cassette subfamily G member 5 |
Gene Aliases: |
STSL |
Location: |
Chr.2:43806154-43838840 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCG5 | NM_022436.2 | NP_071881.1 | ||
| XM_005264480.3 | XP_005264537.1 | |||
| XM_006712073.3 | XP_006712136.1 | |||
| XM_006712074.3 | XP_006712137.1 | |||
| XM_011533024.2 | XP_011531326.1 | |||
| XM_011533025.2 | XP_011531327.1 | |||
| XM_011533026.2 | XP_011531328.1 | |||
| XM_011533027.2 | XP_011531329.1 | |||
| AF312715.2 | AAG53099.1 | |||
| AF320293.1 | AAG40003.1 | |||
| AK091997.1 | ||||
| BC111541.1 | AAI11542.1 |
Target Gene Details
Entrez Gene ID: | 64241 |
Gene Name: | ATP binding cassette subfamily G member 8 |
Gene Aliases: |
GBD4, STSL |
Location: |
Chr.2:43831942-43880051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCG8 | XM_011533029.2 | XP_011531331.1 | ||
| XM_011533030.2 | XP_011531332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012126 | Chr.2:43716882 - 44206414 on Build GRCh38 | Gain |
|
| nsv998912 | Chr.2:43780254 - 43835757 on Build GRCh38 | Gain |
|
| dgv59n68 | Chr.2:43619845 - 43871585 on Build GRCh38 | Gain |
|
| nsv1007740 | Chr.2:42961811 - 44117003 on Build GRCh38 | Gain |
|
| nsv581661 | Chr.2:43762402 - 44145770 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map