Assay Details
Target Gene Details
Entrez Gene ID: | 130132 |
Gene Name: | raftlin family member 2 |
Gene Aliases: |
C2orf11, Raftlin-2 |
Location: |
Chr.2:197568224-197676050 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFTN2 | NM_144629.2 | NP_653230.2 | ||
| XM_011510595.2 | XP_011508897.1 | |||
| XM_011510596.2 | XP_011508898.1 | |||
| XM_011510597.2 | XP_011508899.1 | |||
| XM_011510598.2 | XP_011508900.1 | |||
| XM_017003332.1 | XP_016858821.1 | |||
| AK055136.1 | BAB70861.1 | |||
| BC093966.1 | AAH93966.1 | |||
| BC111069.1 | AAI11070.1 | |||
| BC113361.1 | ||||
| BX647618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014881 | Chr.2:197432432 - 197668882 on Build GRCh38 | Gain |
|
| nsv584156 | Chr.2:197553649 - 197687088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map