Assay Details
Target Gene Details
Entrez Gene ID: | 150709 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
- |
Location: |
Chr.2:189674292-189763059 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKAR | XM_011510673.2 | 1 | 86 | XP_011508975.1 |
| XM_011510674.1 | 1 | 86 | XP_011508976.1 | |
| XM_011510675.2 | 1 | 86 | XP_011508977.1 | |
| XM_011510676.2 | 1 | 86 | XP_011508978.1 | |
| XM_011510677.2 | 1 | 86 | XP_011508979.1 | |
| XM_011510679.2 | 1 | 86 | XP_011508981.1 | |
| XM_011510680.1 | 1 | 86 | XP_011508982.1 | |
| XM_011510681.1 | 1 | 86 | XP_011508983.1 | |
| XM_011510682.2 | 1 | 86 | XP_011508984.1 | |
| XM_011510685.2 | 1 | 86 | XP_011508987.1 | |
| XM_011510686.2 | 1 | 86 | XP_011508988.1 | |
| XM_011510687.2 | 1 | 86 | XP_011508989.1 | |
| XM_011510688.1 | 1 | 86 | XP_011508990.1 | |
| XM_011510689.2 | 1 | 86 | XP_011508991.1 | |
| XM_011510692.2 | 1 | 86 | XP_011508994.1 | |
| XM_011510693.2 | 1 | 86 | XP_011508995.1 | |
| XM_011510694.2 | 1 | 86 | XP_011508996.1 | |
| XM_017003413.1 | 1 | 86 | XP_016858902.1 | |
| XM_017003414.1 | 1 | 86 | XP_016858903.1 | |
| XM_017003415.1 | 1 | 86 | XP_016858904.1 | |
| XM_017003416.1 | 1 | 86 | XP_016858905.1 | |
| XM_017003417.1 | 1 | 86 | XP_016858906.1 | |
| XM_017003418.1 | 1 | 86 | XP_016858907.1 | |
| XM_017003419.1 | 1 | 86 | XP_016858908.1 | |
| XM_017003420.1 | 1 | 86 | XP_016858909.1 | |
| BC044907.1 | 1 | 88 | AAH44907.1 |

Genomic Map