Assay Details
Target Gene Details
Entrez Gene ID: | 130940 |
Gene Name: | coiled-coil domain containing 148 |
Gene Aliases: |
- |
Location: |
Chr.2:158171356-158456753 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC148 | NM_001301684.1 | NP_001288613.1 | ||
| NM_138803.3 | NP_620158.3 | |||
| XM_005246319.3 | XP_005246376.1 | |||
| XM_011510638.2 | XP_011508940.1 | |||
| XM_017003392.1 | XP_016858881.1 | |||
| XM_017003393.1 | XP_016858882.1 | |||
| XM_017003394.1 | XP_016858883.1 | |||
| XM_017003395.1 | XP_016858884.1 | |||
| XM_017003396.1 | XP_016858885.1 | |||
| XM_017003397.1 | XP_016858886.1 | |||
| XM_017003398.1 | XP_016858887.1 | |||
| AF458588.1 | AAM49716.1 | |||
| AK123220.1 | ||||
| BC105928.1 | AAI05929.1 | |||
| BC107596.1 | AAI07597.1 | |||
| BC107597.1 | AAI07598.1 |
Target Gene Details
Entrez Gene ID: | 554201 |
Gene Name: | CCDC148 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:158166650-158236169 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC148-AS1 | NR_038850.1 |
More Information
Set Membership: |
|

Genomic Map