Assay Details
Target Gene Details
Entrez Gene ID: | 3554 |
Gene Name: | interleukin 1 receptor type 1 |
Gene Aliases: |
CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80 |
Location: |
Chr.2:102069638-102179874 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1R1 | NM_001288706.1 | 1 | 81 | NP_001275635.1 |
| NM_001320978.1 | 1 | 81 | NP_001307907.1 | |
| XM_011511115.2 | XP_011509417.1 | |||
| XM_011511117.1 | XP_011509419.1 | |||
| AK314433.1 | 1 | 81 | ||
| DA861783.1 | 1 | 81 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2852 | Chr.2:102062195 - 102100116 on Build GRCh38 | Deletion |
|
| dgv6981n54 | Chr.2:102061304 - 102074850 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map