Assay Details
Target Gene Details
Entrez Gene ID: | 51454 |
Gene Name: | GULP, engulfment adaptor PTB domain containing 1 |
Gene Aliases: |
CED-6, CED6, GULP |
Location: |
Chr.2:188291669-188595926 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Additional Information:
For this assay, SNP(s) [rs72905578] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map