Assay Details
Target Gene Details
Entrez Gene ID: | 140738 |
Gene Name: | transmembrane protein 37 |
Gene Aliases: |
PR, PR1 |
Location: |
Chr.2:119429912-119438520 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM37 | XM_005263598.4 | XP_005263655.1 | ||
| XM_011510659.2 | XP_011508961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592167 | Chr.2:119401097 - 119522758 on Build GRCh38 | Gain |
|
| nsv459119 | Chr.2:118952824 - 119514207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map