Assay Details
Target Gene Details
Entrez Gene ID: | 58538 |
Gene Name: | membrane palmitoylated protein 4 |
Gene Aliases: |
ALS2CR5, DLG6 |
Location: |
Chr.2:201644874-201700263 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP4 | NM_033066.2 | NP_149055.1 | ||
| XM_017004620.1 | XP_016860109.1 | |||
| AB053302.1 | BAB69012.1 | |||
| AB053304.1 | ||||
| AF316032.1 | AAK71862.1 | |||
| AK131208.1 | BAD18399.1 | |||
| AK298589.1 | ||||
| AK298947.1 | ||||
| AK300622.1 | ||||
| BC132785.1 | ||||
| BC144206.1 | ||||
| BM999928.1 | ||||
| BX647162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584274 | Chr.2:201624984 - 201666019 on Build GRCh38 | Loss |
|
| dgv7249n54 | Chr.2:201644883 - 201666824 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112212386] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map