Assay Details
Target Gene Details
Entrez Gene ID: | 9167 |
Gene Name: | cytochrome c oxidase subunit 7A2 like |
Gene Aliases: |
COX7AR, COX7RP, EB1, SIG81 |
Location: |
Chr.2:42335561-42368957 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX7A2L | NM_001319038.1 | NP_001305967.1 | ||
| CA336587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583757 | Chr.2:42335681 - 42347279 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77941865] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map