Assay Details
Target Gene Details
Entrez Gene ID: | 165186 |
Gene Name: | family with sequence similarity 179 member A |
Gene Aliases: |
- |
Location: |
Chr.2:28981298-29052230 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM179A | NM_001321538.1 | NP_001308467.1 | ||
| NM_001321539.1 | NP_001308468.1 | |||
| NM_199280.2 | NP_954974.2 | |||
| XM_011532628.2 | XP_011530930.1 | |||
| XM_017003512.1 | XP_016859001.1 | |||
| AK296977.1 | ||||
| BI768759.1 | ||||
| DA213268.1 | ||||
| DA460119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34904 | Chr.2:28979483 - 30284483 on Build GRCh38 | Gain |
|
| esv3590167 | Chr.2:28971589 - 29053949 on Build GRCh38 | Gain |
|
| nsv1008490 | Chr.2:28884768 - 29482018 on Build GRCh38 | Gain |
|
| esv2759035 | Chr.2:28953630 - 30316563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map