Assay Details
Target Gene Details
Entrez Gene ID: | 10791 |
Gene Name: | vesicle associated membrane protein 5 |
Gene Aliases: |
- |
Location: |
Chr.2:85584408-85593388 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAMP5 | NM_006634.2 | NP_006625.1 | ||
| AF054825.1 | AAC97473.1 | |||
| AF151025.1 | AAF36111.1 | |||
| AK292660.1 | ||||
| BC017891.1 | AAH17891.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114148288] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map