Assay Details
Target Gene Details
Entrez Gene ID: | 100506054 |
Gene Name: | RNASEH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:3558386-3561750 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH1-AS1 | NR_038429.1 | |||
| NR_038430.1 | ||||
| NR_038431.1 | ||||
| AK023018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819972 | Chr.2:3551407 - 3560402 on Build GRCh38 | Gain |
|
| nsv1011656 | Chr.2:2931099 - 3576389 on Build GRCh38 | Gain |
|
| dgv3702n100 | Chr.2:3361936 - 3733306 on Build GRCh38 | Gain |
|
| nsv580822 | Chr.2:3552110 - 4305551 on Build GRCh38 | Gain |
|

Genomic Map