Assay Details
Target Gene Details
Entrez Gene ID: | 57404 |
Gene Name: | cytochrome P450 family 20 subfamily A member 1 |
Gene Aliases: |
CYP-M |
Location: |
Chr.2:203238441-203305840 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP20A1 | NM_177538.2 | NP_803882.1 | ||
| XM_005246698.3 | XP_005246755.1 | |||
| XM_005246699.3 | XP_005246756.1 | |||
| XM_005246701.3 | XP_005246758.1 | |||
| XM_006712644.3 | XP_006712707.1 | |||
| XM_006712645.3 | XP_006712708.1 | |||
| XM_017004555.1 | XP_016860044.1 | |||
| AF183412.1 | AAG09681.1 | |||
| AK297082.1 | ||||
| AK307931.1 | ||||
| AL832998.1 | CAH56305.1 | |||
| AY359068.1 | AAQ89427.1 | |||
| BC020616.1 | AAH20616.1 | |||
| BC093022.1 | AAH93022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map