Assay Details
Target Gene Details
Entrez Gene ID: | 100506299 |
Gene Name: | ID2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.2:8670984-8681863 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ID2-AS1 | NR_110153.1 | 2 | 659 | |
| NR_110154.1 | ||||
| AA970710.1 | 1 | 168 | ||
| AI022540.1 | ||||
| AI034463.1 | 1 | 168 | ||
| DA317685.1 | ||||
| DB336412.1 | 1 | 150 |

Genomic Map