Assay Details
Target Gene Details
Entrez Gene ID: | 3554 |
Gene Name: | interleukin 1 receptor type 1 |
Gene Aliases: |
CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80 |
Location: |
Chr.2:102069638-102179874 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1R1 | NM_001288706.1 | NP_001275635.1 | ||
| NM_001320978.1 | NP_001307907.1 | |||
| XM_011511115.2 | XP_011509417.1 | |||
| XM_011511117.1 | XP_011509419.1 | |||
| AK314433.1 | ||||
| DA861783.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs151010796] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map