Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Gene Details
Entrez Gene ID: | 11262 |
Gene Name: | SP140 nuclear body protein |
Gene Aliases: |
LYSP100, LYSP100-A, LYSP100-B |
Location: |
Chr.2:230202924-230316571 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP140 | XM_011510517.2 | XP_011508819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584666 | Chr.2:230185999 - 230241906 on Build GRCh38 | Gain |
|
| esv29172 | Chr.2:230208263 - 230603132 on Build GRCh38 | Loss |
|
| nsv998291 | Chr.2:230180672 - 230271563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map