Assay Details
Target Gene Details
Entrez Gene ID: | 339745 |
Gene Name: | speckle type BTB/POZ protein like |
Gene Aliases: |
BTBD33 |
Location: |
Chr.2:138501780-138573235 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPOPL | NM_001001664.2 | NP_001001664.1 | ||
| XM_005263655.4 | XP_005263712.1 | |||
| XM_005263656.4 | XP_005263713.1 | |||
| XM_005263657.4 | XP_005263714.1 | |||
| BC071613.1 | AAH71613.1 | |||
| CN315400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834390 | Chr.2:138411846 - 138576459 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map