Assay Details
Target Gene Details
Entrez Gene ID: | 100630918 |
Gene Name: | promoter of MAT2A antisense radiation-induced circulating long non-coding RNA |
Gene Aliases: |
PARTICLE |
Location: |
Chr.2:85537467-85538886 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARTICL | NR_038942.1 | 1 | 681 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2811 | Chr.2:85535833 - 85563891 on Build GRCh38 | Insertion |
|

Genomic Map