Assay Details
Target Gene Details
Entrez Gene ID: | 51057 |
Gene Name: | WD repeat containing planar cell polarity effector |
Gene Aliases: |
BBS15, C2orf86, CHDTHP, FRITZ, FRTZ |
Location: |
Chr.2:63120755-63840825 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDPCP | NM_001042692.2 | NP_001036157.1 | ||
| NM_015910.5 | NP_056994.3 | |||
| NR_122106.1 | ||||
| XM_005264348.3 | XP_005264405.1 | |||
| XM_011532881.2 | XP_011531183.1 | |||
| XM_017004251.1 | XP_016859740.1 | |||
| XM_017004252.1 | XP_016859741.1 | |||
| AF131737.1 | AAD20026.1 | |||
| AK001849.1 | ||||
| AK022647.1 | ||||
| AL833623.1 | ||||
| BC093752.1 | AAH93752.1 | |||
| BC093754.1 | AAH93754.1 | |||
| BX647132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004678 | Chr.2:63003587 - 63335349 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map