Assay Details
Target Gene Details
Entrez Gene ID: | 7844 |
Gene Name: | ring finger protein 103 |
Gene Aliases: |
HKF-1, KF-1, KF1, ZFP-103, ZFP103 |
Location: |
Chr.2:86603393-86623877 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF103 | NM_001198951.1 | NP_001185880.1 | ||
| NM_005667.3 | NP_005658.1 | |||
| AK122978.1 | 1 | 462 | ||
| AK314180.1 | ||||
| BC035053.2 | AAH35053.1 | |||
| BC110333.1 | AAI10334.1 | |||
| D76444.1 | BAA19739.1 | |||
| DA816879.1 |
Target Gene Details
Entrez Gene ID: | 100526767 |
Gene Name: | RNF103-CHMP3 readthrough |
Gene Aliases: |
RNF103-VPS24 |
Location: |
Chr.2:86503430-86721122 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF103-CHMP3 | NM_001198954.1 | NP_001185883.1 | ||
| AK294389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011590 | Chr.2:86597058 - 86678991 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map