Assay Details
Target Gene Details
Entrez Gene ID: | 58190 |
Gene Name: | CTD small phosphatase 1 |
Gene Aliases: |
NIF3, NLI-IF, NLIIF, SCP1 |
Location: |
Chr.2:218398338-218405941 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTDSP1 | NM_001206878.1 | NP_001193807.1 | ||
| NM_021198.2 | NP_067021.1 | |||
| NM_182642.2 | NP_872580.1 | |||
| XM_011511569.2 | XP_011509871.1 | |||
| XM_011511570.2 | XP_011509872.1 | |||
| XM_017004615.1 | XP_016860104.1 | |||
| XM_017004616.1 | XP_016860105.1 | |||
| XM_017004617.1 | XP_016860106.1 | |||
| XM_017004618.1 | XP_016860107.1 | |||
| AF229162.1 | AAG15402.1 | |||
| AY279529.1 | AAP34397.1 | |||
| BC012977.2 | AAH12977.1 | |||
| BX424971.2 | ||||
| BX446444.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519829 | Chr.2:218286495 - 218403058 on Build GRCh38 | Loss |
|
| nsv3166 | Chr.2:218371765 - 218405183 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74435244,rs79924237] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map