Assay Details
Target Gene Details
Entrez Gene ID: | 80122 |
Gene Name: | mitogen-activated protein kinase kinase kinase 19 |
Gene Aliases: |
RCK, YSK4 |
Location: |
Chr.2:134963875-135047468 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K19 | NM_001018044.2 | NP_001018054.1 | ||
| NM_001018046.2 | NP_001018056.1 | |||
| NM_001018047.2 | NP_001018057.1 | |||
| NM_001282883.1 | NP_001269812.1 | |||
| NM_025052.4 | NP_079328.3 | |||
| XM_011511891.2 | XP_011510193.1 | |||
| XM_011511892.1 | XP_011510194.1 | |||
| XM_011511894.2 | XP_011510196.1 | |||
| XM_011511896.2 | XP_011510198.1 | |||
| XM_011511897.2 | XP_011510199.1 | |||
| XM_017005003.1 | XP_016860492.1 | |||
| XM_017005004.1 | XP_016860493.1 | |||
| XM_017005005.1 | XP_016860494.1 | |||
| XM_017005006.1 | XP_016860495.1 | |||
| AY574901.1 | AAT81410.1 | |||
| AY574902.1 | AAT81411.1 | |||
| AY574903.1 | AAT81412.1 | |||
| AY574904.1 | AAT81413.1 | |||
| AY574905.1 | AAT81414.1 | |||
| BC137276.1 | ||||
| BC137277.1 | ||||
| BC144555.1 |
More Information
Set Membership: |
|

Genomic Map