Assay Details
Target Gene Details
Entrez Gene ID: | 51665 |
Gene Name: | ankyrin repeat and SOCS box containing 1 |
Gene Aliases: |
ASB-1 |
Location: |
Chr.2:238426741-238452250 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB1 | NM_001040445.1 | NP_001035535.1 | ||
| XM_005246080.1 | XP_005246137.1 | |||
| AF156777.1 | AAD41894.1 | |||
| BC014528.2 | AAH14528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map