Assay Details
Target Gene Details
Entrez Gene ID: | 653140 |
Gene Name: | family with sequence similarity 228 member A |
Gene Aliases: |
C2orf84 |
Location: |
Chr.2:24175043-24191698 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM228A | NM_001040710.2 | NP_001035800.1 | ||
| BC050462.1 | AAH50462.1 | |||
| HY022970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv581179 | Chr.2:24114991 - 24317722 on Build GRCh38 | Gain |
|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142051385] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map