Assay Details
Target Gene Details
Entrez Gene ID: | 4735 |
Gene Name: | septin 2 |
Gene Aliases: |
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5 |
Location: |
Chr.2:241315187-241354027 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
More Information
Set Membership: |
|

Genomic Map