Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP110 | NM_001185015.1 | NP_001171944.1 | ||
| NM_004509.3 | NP_004500.3 | |||
| NM_004510.3 | NP_004501.3 | |||
| NM_080424.2 | NP_536349.2 | |||
| XM_005246525.3 | XP_005246582.1 | |||
| XM_006712487.3 | XP_006712550.1 | |||
| XM_006712489.3 | XP_006712552.1 | |||
| XM_011511088.2 | XP_011509390.1 | |||
| XM_011511089.2 | XP_011509391.1 | |||
| XM_011511090.2 | XP_011509392.1 | |||
| XM_011511091.2 | XP_011509393.1 | |||
| XM_011511092.2 | XP_011509394.1 | |||
| XM_017003968.1 | XP_016859457.1 | |||
| XM_017003969.1 | XP_016859458.1 | |||
| XM_017003970.1 | XP_016859459.1 | |||
| AF280094.1 | AAF99318.1 | |||
| AF280095.1 | AAG09826.1 | |||
| AK026488.1 | ||||
| AK301097.1 | ||||
| BC012447.2 | AAH12447.1 | |||
| BC019059.2 | AAH19059.1 | |||
| DA958469.1 | ||||
| DB474530.1 |
Target Gene Details
Entrez Gene ID: | 11262 |
Gene Name: | SP140 nuclear body protein |
Gene Aliases: |
LYSP100, LYSP100-A, LYSP100-B |
Location: |
Chr.2:230202924-230316571 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP140 | XM_011510517.2 | XP_011508819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584666 | Chr.2:230185999 - 230241906 on Build GRCh38 | Gain |
|
| esv29172 | Chr.2:230208263 - 230603132 on Build GRCh38 | Loss |
|
| nsv998291 | Chr.2:230180672 - 230271563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map