Assay Details
Target Gene Details
Entrez Gene ID: | 84620 |
Gene Name: | ST6 beta-galactoside alpha-2,6-sialyltransferase 2 |
Gene Aliases: |
SIAT2, ST6GalII |
Location: |
Chr.2:106801600-106887410 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GAL2 | NM_001142351.1 | NP_001135823.1 | ||
| NM_001142352.1 | NP_001135824.1 | |||
| NM_001322362.1 | NP_001309291.1 | |||
| NM_032528.2 | NP_115917.1 | |||
| NR_136313.1 | ||||
| XM_006712802.1 | XP_006712865.1 | |||
| XM_011511999.2 | XP_011510301.1 | |||
| XM_011512000.2 | XP_011510302.1 | |||
| XM_011512001.2 | XP_011510303.1 | |||
| XM_017005110.1 | XP_016860599.1 | |||
| AB058780.1 | ||||
| AB059555.1 | BAC24793.1 | |||
| AJ512141.1 | CAD54408.1 | |||
| AK307784.1 | ||||
| BC008680.1 | AAH08680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6988n54 | Chr.2:106262145 - 107828618 on Build GRCh38 | Loss |
|
| nsv471617 | Chr.2:106579606 - 107474705 on Build GRCh38 | Gain |
|
| nsv1014939 | Chr.2:106771852 - 106883422 on Build GRCh38 | Loss |
|
| esv2720482 | Chr.2:106245437 - 107913028 on Build GRCh38 | Deletion |
|
| dgv4034n100 | Chr.2:106236698 - 108333964 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map