Assay Details
Target Gene Details
Entrez Gene ID: | 27179 |
Gene Name: | interleukin 36, alpha |
Gene Aliases: |
FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6 |
Location: |
Chr.2:113005461-113011071 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL36A | XM_011510965.1 | XP_011509267.1 |
More Information
Set Membership: |
|

Genomic Map