Assay Details
Target Gene Details
Entrez Gene ID: | 1593 |
Gene Name: | cytochrome P450 family 27 subfamily A member 1 |
Gene Aliases: |
CP27, CTX, CYP27 |
Location: |
Chr.2:218781006-218815293 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP27A1 | XM_017003488.1 | 1 | 578 | XP_016858977.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518750 | Chr.2:218751245 - 218804652 on Build GRCh38 | Loss |
|
| esv3594349 | Chr.2:218756892 - 218789452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map