Assay Details
Target Gene Details
Entrez Gene ID: | 254042 |
Gene Name: | methionyl aminopeptidase type 1D (mitochondrial) |
Gene Aliases: |
MAP 1D, MAP1D, MetAP 1D, Metap1l |
Location: |
Chr.2:171999897-172082430 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METAP1D | NM_001322278.1 | NP_001309207.1 | ||
| NM_001322279.1 | NP_001309208.1 | |||
| NM_199227.2 | NP_954697.1 | |||
| NR_136273.1 | ||||
| NR_136276.1 | ||||
| AY374142.1 | AAR27795.1 | |||
| BC113644.1 | ||||
| DQ005576.1 |
More Information
Set Membership: |
|

Genomic Map