Assay Details
Target Gene Details
Entrez Gene ID: | 23620 |
Gene Name: | neurotensin receptor 2 |
Gene Aliases: |
NTR2 |
Location: |
Chr.2:11658178-11670206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTSR2 | NM_012344.3 | NP_036476.1 | ||
| XM_005246156.3 | XP_005246213.1 | |||
| XM_006711876.3 | XP_006711939.1 | |||
| XM_006711877.3 | XP_006711940.1 | |||
| XM_017003738.1 | XP_016859227.1 | |||
| AY429107.1 | AAR07902.1 | |||
| BC022501.1 | AAH22501.1 | |||
| BC037776.2 | AAH37776.1 | |||
| BI758259.1 | ||||
| Y10148.1 | CAA71233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580972 | Chr.2:11668528 - 11713886 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79333363] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map