Assay Details
Target Gene Details
Entrez Gene ID: | 11227 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 5 |
Gene Aliases: |
GALNAC-T5, GALNACT5 |
Location: |
Chr.2:157257548-157318491 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT5 | NM_014568.1 | NP_055383.1 | ||
| XM_017003237.1 | XP_016858726.1 | |||
| XM_017003238.1 | XP_016858727.1 | |||
| AF154107.1 | AAF15313.1 | |||
| AJ245539.1 | CAB65104.1 | |||
| AK292154.1 | ||||
| AY277591.1 | AAP34404.1 | |||
| BC142676.1 | ||||
| BC142703.1 | 1 | 3615 |
More Information
Additional Information:
For this assay, SNP(s) [rs74443166] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map