Assay Details
Target Gene Details
Entrez Gene ID: | 98 |
Gene Name: | acylphosphatase 2 |
Gene Aliases: |
ACYM, ACYP |
Location: |
Chr.2:53970838-54305300 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACYP2 | NM_001320586.1 | NP_001307515.1 | ||
| NM_001320587.1 | NP_001307516.1 | |||
| XM_017005406.1 | XP_016860895.1 | |||
| XM_017005407.1 | XP_016860896.1 | |||
| XM_017005409.1 | XP_016860898.1 | |||
| XM_017005411.1 | XP_016860900.1 | |||
| XM_017005412.1 | XP_016860901.1 | |||
| XM_017005413.1 | XP_016860902.1 | |||
| BG741807.1 | ||||
| BU543943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2720078 | Chr.2:53962225 - 55281329 on Build GRCh38 | Deletion |
|
| nsv1001246 | Chr.2:53968517 - 54207695 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map