Assay Details
Target Gene Details
Entrez Gene ID: | 7850 |
Gene Name: | interleukin 1 receptor type 2 |
Gene Aliases: |
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB |
Location: |
Chr.2:101991805-102028544 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1R2 | NM_001261419.1 | NP_001248348.1 | ||
| NM_004633.3 | NP_004624.1 | |||
| XM_006712734.3 | XP_006712797.1 | |||
| XM_006712736.3 | XP_006712799.1 | |||
| XM_017004889.1 | XP_016860378.1 | |||
| AK130412.1 | ||||
| AK309161.1 | ||||
| BC039031.1 | AAH39031.1 |
More Information
Set Membership: |
|

Genomic Map