Assay Details
Target Gene Details
Entrez Gene ID: | 5495 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1B |
Gene Aliases: |
PP2C-beta, PP2C-beta-X, PP2CB, PP2CBETA, PPC2BETAX |
Location: |
Chr.2:44168803-44247330 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1B | NM_001033557.2 | NP_001028729.1 | ||
| NM_002706.5 | NP_002697.1 | |||
| NM_177968.3 | NP_808907.1 | |||
| NM_177969.3 | NP_808908.1 | |||
| XM_011532936.2 | XP_011531238.1 | |||
| XM_017004395.1 | XP_016859884.1 | |||
| XM_017004396.1 | XP_016859885.1 | |||
| XM_017004397.1 | XP_016859886.1 | |||
| AJ271832.1 | CAC27992.1 | |||
| AJ271835.1 | CAC27993.1 | |||
| AK056009.1 | ||||
| AK291214.1 | ||||
| AL833035.1 | CAH56319.1 | |||
| BC012002.1 | AAH12002.1 | |||
| BC064381.1 | AAH64381.1 | |||
| BF979498.1 | ||||
| DA128532.1 | ||||
| DQ023508.1 | AAY89639.1 | |||
| DQ023509.1 | AAY89640.1 | |||
| DQ023510.1 | AAY89641.1 | |||
| DQ023511.1 | AAY89642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012126 | Chr.2:43716882 - 44206414 on Build GRCh38 | Gain |
|
| nsv1006405 | Chr.2:44013164 - 44709137 on Build GRCh38 | Loss |
|
| nsv953775 | Chr.2:44169262 - 44187261 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map